grade
Molecular Biology
for molecular biology
form
buffered aqueous solution
shipped in
dry ice
storage temp.
−20°C
General description
The kit provides expression vectors ”pFLAG-CMV”-2 and pc-Myc-CMV-2 designed for the cloning and expression of protein interaction candidates as N-terminal FLAG and N-terminal c-Myc fusions. Control plasmids pFLAG-CMV-2-p53, pC-Myc-CMV-2-Large T antigen and pC-Myc-CMV-2-BAP supplied are for expression of positive and negative binding partners in the IP analysis. The kit also includes the MF-2 and MR-2 verification primers.
Vector Maps and Sequences
Features and Benefits
- Fast and convenient 96 well plate format
- Cleaner analysis due to strongly bound antibody
- Rapid, sensitive, quantitative results
Components
- pFLAG-CMV-2 Expression Vector (10 μg)
- pc-Myc-CMV-2 Expression Vector (10 μg)
- pFLAG-CMV-2-p53 control plasmid (20 μg)
- pc-Myc-CMV-2-Large T antigen control plasmid (10 μg)
- pc-Myc-CMV-2-BAP control plasmid (10 μg)
- Verification Primer-MF (2 nmol)
- Verification Primer-MR (2 nmol)
Kit Components Only
- pc-Myc-CMV-2 10 μg
- pc-Myc-CMV-2-BAP 10 μg
- pc-Myc-CMV-2-Large T antigen 10 μg
- pFLAG-CMV™-2 10 μg
- pFLAG-CMV™-2-p53 20 μg
- Verification Primer-MF 2 nmol
- Verification Primer-MR 2 nmol
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service