Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS7724T30

PYREX® PTFE pestle tissue grinder with steel shaft

capacity 30 mL

Synonym(s):

homogenizer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
Product CLS7724T30 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Corning

capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

PYREX is a registered trademark of Corning, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service