CLS6767
Corning® LSE™ mini microcentrifuges
8 x 1.5/2.0 mL rotor and 4-place strip tube rotors included, 230V, UK plug, 1/cs
Synonym(s):
centrifuges, microcentrifuges, microfuge
About This Item
Recommended Products
AC/DC input
230 V AC, UK plug
description
8-place rotor that will accept standard 1.5 mL to 2.0 mL microcentrifuge tubes, and adapters are included for 0.5/0.4 mL and 0.2 mL tubes. Also included is a 4-place rotor for PCR strip tubes.
feature
CE compliant
to hold 4 PCR strip tubes
to hold 8 1.5/2.0 mL microcentrifuge tubes
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 6767
parameter
2000 × g speed
2000 × g
6000 rpm speed
6000 rpm
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
- Includes an 8-place rotor that will accept standard 1.5 mL to 2.0 mL microcentrifuge tubes
- Includes adapters for 0.5/0.4 mL and 0.2 mL tubes
- 4-place rotor for PCR strip tubes included
- Compact, lightweight and easy-to-use
- Close the lid to start, press the lid release button to stop
- 8 x 1.5/2.0 mL rotor and 4-place strip tube rotors included
- Fixed speed of 6000 rpm generates 2000 x g
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service