Key Documents
CLS4888
Corning® Multipette® adjustable volume multichannel pipettor
8-pette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.78
feature
8-channel
electronic: no
manufacturer/tradename
Corning 4888
volume measuring range
20-200 μL
volume
200 μL
General description
The 8-Pette and 12-Pette offer features for ease of use, versatility, and safety:
  • 20-200 μL per channel in 0.5 μL increments
  • Fully autoclavable
  • Pistol grip eliminates thumb fatigue
  • Separate tip ejector prevents accidental ejection
  • Any number of channels can be left un-used without affecting accuracy
  • Horizontal configuration moves hand closer to tips for better control of placement
Features and Benefits
1-8 channel pipette
Legal Information
Corning is a registered trademark of Corning, Inc.
Multipette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Take a deep dive into the world of 3D cell culture courtesy of Corning and understand why the Corning® spheroid microplate can be an asset in the evolution of 3D cell culture.
Protocols
Understand how Corning® Spheroid Microplates can be used for generating and culturing tumor spheroids in a 96-well ­spheroid microplate format.