CLS4880
Corning® Multipette® adjustable volume multichannel pipettor
12-pette
About This Item
Recommended Products
feature
12-channel
electronic: no
manufacturer/tradename
Corning 4880
volume measuring range
20-200 μL
volume
200 μL
General description
- 20-200 μL per channel in 0.5 μL increments
- Fully autoclavable
- Pistol grip eliminates thumb fatigue
- Separate tip ejector prevents accidental ejection
- Any number of channels can be left un-used without affecting accuracy
- Horizontal configuration moves hand closer to tips for better control of placement
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
If you need assistance, please contact Customer Support
Already Own This Product?
Articles
Take a deep dive into the world of 3D cell culture courtesy of Corning and understand why the Corning® spheroid microplate can be an asset in the evolution of 3D cell culture.
Protocols
Understand how Corning® Spheroid Microplates can be used for generating and culturing tumor spheroids in a 96-well spheroid microplate format.