CLS4519161
Corning® ProCulture® spinner flask accessories
Gas and Media Handling Fitting with Dual 1/8 in. Inlets (1 long, 1 short tube) for 8L Angled Sidearm Flasks
Synonym(s):
container accessory
Select a Size
$402.00
Available to ship onMay 05, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$402.00
Available to ship onMay 05, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Recommended Products
manufacturer/tradename
Corning 4519-161
Related Categories
General description
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)