CLS4519126
Corning® ProCulture® spinner flask accessories
Media Handling Fitting with Single 1/2 in. Inlets for 36L Vertical Sidearm Flasks
Synonym(s):
container accessory
Select a Size
$422.00
Available to ship onMay 01, 2025Details
Select a Size
About This Item
$422.00
Available to ship onMay 01, 2025Details
manufacturer/tradename
Corning 4519-126
Related Categories
General description
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)