Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS430289

Corning® cryogenic vial, external thread with plug seal cap

2 mL external threaded polypropylene cryogenic vial w/ round bottom and plug seal cap, 500/cs

Synonym(s):

cryogenic tubes, cryogenic vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

natural polypropylene vial
orange polypropylene cap

sterility

sterile; γ-irradiated (certified nonpyrogenic)

feature

self-standing: no
round bottom
silicone washer: no

packaging

pack of 50
case of 500

manufacturer/tradename

Corning 430289

capacity

2.0 mL

General description

This Corning vial is manufactured from polypropylene to withstand temperatures to -196°C. Vial features an external thread with the traditional plug-seal cap design for a secure seal.

  • 2 mL capacity with round bottom
  • Manufactured from polypropylene to withstand temperatures to -196°C
  • Plug seal cap for a secure seal
  • Sterilized by gamma radiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service