Key Documents
CLS401023
Corning® Power Supply for Slow Speed, Submersible and Multiple Position Stirrers
AC/DC input 120 V AC
Synonym(s):
stirrer power supply
Select a Size
1 EA
$225.00
$225.00
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
1 EA
$225.00
About This Item
UNSPSC Code:
41103800
$225.00
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Related Categories
General description
This is a replacement power pack for Corning Slow-Speed Stirrers (SS2L, SS10L, SS4R, SS4L and SS10R), Submersible Stirrers (IM75 and IM150) and Multiple Position Stirrers (MP4L, MP5L, MP9L and MP4R). It operates on 120V/60Hz and is UL approved to meet needs in North America and other locations with similar requirements.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)