CLS40082
Corning® microarray slide mailers/storage boxes
5 slide mailer
Synonym(s):
Corning® 40083
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123302
material
polycarbonate
feature
Holds 5 x slides 76x26 mm
packaging
pack of 50 ea
manufacturer/tradename
Corning 40082
color
orange
Looking for similar products? Visit Product Comparison Guide
General description
The plastic containers in which UltraGAP Coated Slides are shipped also function as storage boxes for printed arrays. These containers are available as either 5 slide mailers or 25 slide storage boxes. The trans-orange plastic has low transmittance in the 500 to 700 nm wavelength range which helps protect Cy3 and Cy5 dyes from photobleaching. These rigid plastic containers do not shed particles or outgas volatile chemicals that may contaminate microarray slides.
The Corning 25 slider storage box has a lift off lid which is easy to open and close. The 5 slide mailer has a hinged lid that snaps closed tightly to prevent slides from accidentlly falling out.
The Corning 25 slider storage box has a lift off lid which is easy to open and close. The 5 slide mailer has a hinged lid that snaps closed tightly to prevent slides from accidentlly falling out.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service