Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS40081

Corning® microarray slide mailers/storage boxes

25 slide storage box, orange

Synonym(s):

Corning® 40083

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123302

material

polycarbonate

feature

Holds 25 x slides 76x26 mm

packaging

pack of 20 ea

manufacturer/tradename

Corning 40081

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

The plastic containers in which UltraGAP Coated Slides are shipped also function as storage boxes for printed arrays. These containers are available as either 5 slide mailers or 25 slide storage boxes. The trans-orange plastic has low transmittance in the 500 to 700 nm wavelength range which helps protect Cy3 and Cy5 dyes from photobleaching. These rigid plastic containers do not shed particles or outgas volatile chemicals that may contaminate microarray slides.
The Corning 25 slider storage box has a lift off lid which is easy to open and close. The 5 slide mailer has a hinged lid that snaps closed tightly to prevent slides from accidentlly falling out.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service