Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3988

Corning® Osteo Assay Surface Multiple Well Plate

size 96 well, flat bottom clear, pkg of (individually wrapped), pkg of (4/case), sterile, lid

Synonym(s):

96 well cell culture plates, cell culture plates, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

clear bottom
flat bottom clear

sterility

sterile

feature

lid
skirt
plate format: 96 well standard

manufacturer/tradename

Corning 3988

packaging

pkg of (4/case)
pkg of (individually wrapped)

size

96 well

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

well working volume

75-200 μL

wells

96 wells

color

clear

suitability

suitable for (Osteoclast and Osteoblast differentiation and function studies)

binding type

(Osteo Assay Surface)

Looking for similar products? Visit Product Comparison Guide

General description

  • Bone biomimetic synthetic surface
  • Inorganic, non-biological
  • Compatible with osteoblast and osteoclast differentiation and functional studies
  • Co-culture of ostoeclast and osteoblast with other cell lines
  • 96 well format uniformly coated
  • Flat bottoms
  • Recommended working volume 75 to 200 μL
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumeric codes for well identification
  • •ndividually wrapped
  • Sterilized by gamma irradiation
Corning Osteo Assay Surface multiple well plates can be used to assess cell functional activity in response to drug treatment. Research related to basic osteoblast biology focuses on cell differentiation and function. Osteoblasts are of mesenchymal origin. Several growth factors have been implicated in osteoblast differentiation, including bone morphogenic proteins (BMPs) and Indian hedgehog. Additionally, several hormones are believed to play an important role in bone formation, including estradiol, parathyroid hormone and leptin. Osteoblast differentiation on the new Corning Osteo Assay Surface provides an in vivo-like surface for improved osteoblast growth and differentiation.

Understanding the biology of bone disease is of critical importance to the aging population. Due to the difficult nature of the assays involved in studying bone, scientists have long been limited with regard to the methods, available formats and throughput used in this area of research. Corning Life Sciences has developed a new platform to aid researchers with their bone studies that is consistent and repeatable. The Corning Osteo Assay Surface multiple well plates are coated with a proprietary synthetic inorganic bone mimetic, which allows the direct assessment of osteoclast and osteoblast activity in vitro. This surface permits easy and reliable quantification of results in a format that is amenable to drug screening, as well as basic research applications.

Osteo Surface

  • Assess cell functionality in response to drug treatment
  • in vivo-like surface for osteoblast growth and differentiation
  • Synthetic inorganic bone mimetic surface
  • Direct assessment of osteoclast and osteoblast activity in vitro
  • Surface amenable to drug screening and other basic research applications

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service