Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3978

Corning® Synthemax®-R surface multiwell plates

6 wells, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

clear wells
polystyrene
round wells

sterility

sterile

feature

lid

packaging

pack of 2 ea

manufacturer/tradename

Corning 3978

size

6 wells

well surface area

9.5 cm2

well volume

16.8 mL

well working volume

16.8 mL

color

clear

suitability

suitable for (the differentiation of stem cells into specialized cell types)

Looking for similar products? Visit Product Comparison Guide

General description

Corning Synthemax cell culture surface uses an active peptide acrylate for polystyrene binding to promote stem cell proliferation and differentiation.

Corning Synthemax-R Surface is a unique synthetic surface coated onto tissue culture treated cell culture flasks and multiwell plates. The surface chemistry is designed to mimic a cells′ natural environment with an extracellular matrix derived cell adhesion promoting peptide. The peptide acrylate coating creates a uniform active surface for stem cell attachment, growth and differentiation, especially in chemically defined media.

Synthemax-R Surface multiple well plates and flasks offer a synthetic alternative to biological coatings and/or feeder cell layers used in traditional stem cell culture. The proprietary surface coating technology ensures lot-to-lot consistency, facilitating reproducible results in laboratories.

Developed by Corning scientists, this new biomimetic surface benefits are:
  • Unique synthetic surface mimics natural cell environment
  • Supports expansion of undifferentiated stem cells into a specialized, differentiated cell type
  • Ready to use surface
  • 6 well format uniformly coated
  • Recommended working volume 1.9 mL to 2.9 mL
  • Nonreversible lids with condensation rings to reduce contamination
  • Individually wrapped
  • Sterilized by gamma irradiation (SAL 10-3)
  • 2 year shelf life
  • Amenable to automation
  • Eliminates the need and time for expensive coating

Legal Information

Corning is a registered trademark of Corning, Inc.
Synthemax is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service