Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3972

Corning® Synthemax®-T surface 75 cm2 flask

rectangular canted neck cell culture flask with vented cap, sterile

Synonym(s):

tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

canted neck
polystyrene

sterility

sterile

feature

closure type Vented cap
graduations
lid

packaging

pack of 2 ea

manufacturer/tradename

Corning 3972

surface area

75 cm2

working volume

15-22.5 μL , 60 mL maximum

color

clear

suitability

suitable for (stem cell proliferation and differentiation)

Looking for similar products? Visit Product Comparison Guide

General description

Corning Synthemax cell culture surface uses an active peptide acrylate for polystyrene binding to promote stem cell proliferation and differentiation.


The Corning Synthemax Surface is a unique synthetic surface coated onto multiwell plates and flasks that mimics the natural cell environment. This surface supports the expansion of stem cells in their undifferentiated state and enables directed differentiation into specialized cell types.

  • Synthemax surface multiple well plate
  • Unique synthetic surface mimics natural cell environment
  • Supports expansion of undifferentiated stem cells
  • Ready to use surface
  • Manufactured from optically clear virgin polystyrene
  • Quality tested for lot-to-lot consistency
  • Recommended working volume 0.2 - 0.3 mL/cm2
  • Individually wrapped
  • Sterilized by gamma irradiation (SAL 10-6)

Legal Information

Corning is a registered trademark of Corning, Inc.
Synthemax is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service