Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3876

Corning® Synthemax®-T surface multiwell plates

6 wells, sterile

Synonym(s):

tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear wells
polystyrene
round wells
size:
6 wells

material

clear wells
polystyrene
round wells

sterility

sterile

feature

lid

packaging

pack of 2 ea

manufacturer/tradename

Corning 3876

size

6 wells

well surface area

9.5 cm2

well volume

16.8 mL

well working volume

1.9-2.9 μL
16.8 mL

color

clear

suitability

suitable for (the differentiation of stem cells into specialized cell types)

Looking for similar products? Visit Product Comparison Guide

General description

Corning Synthemax cell culture surface uses an active peptide acrylate for polystyrene binding to promote stem cell proliferation and differentiation.

The Corning Synthemax Surface is a unique synthetic surface coated onto multiple well plates that mimics the natural cell environment. This surface supports the expansion of stem cells in their undifferentiated state and enables directed differentiation into specialized cell types.

  • Unique synthetic surface mimics natural cell environment
  • Supports expansion of undifferentiated stem cells
  • 6 well format uniformly coated
  • Recommended working volume 1.9 mL to 2.9 mL
  • Nonreversible lids with condensation rings to reduce contamination

Legal Information

Corning is a registered trademark of Corning, Inc.
Synthemax is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service