Key Documents
CLS3845
Corning® 384 well plates
flat bottom clear polystyrene wells, lid, sterile
About This Item
UNSPSC Code:
41100000
material:
black polystyrene plate
flat bottom clear polystyrene wells
polystyrene
size:
384 wells
material
black polystyrene plate
flat bottom clear polystyrene wells
polystyrene
sterility
sterile
feature
lid
packaging
case of 100
manufacturer/tradename
Corning 3845
maximum volume
95 μL
size
384 wells
working volume
80 μL
color
clear
suitability
suitable for (cell cuture)
binding type
poly-D-lysine coated surface
General description
  • Black wall plates have low background fluorescence, minimal light scatter and reduced cross-talk
  • Opaque walls to prevent well-to-well cross-talk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 112μL total volume
  • Recommended working volumes of 20 to 80μL
  • Poly-D-lysine coated for enhanced cell attachment
  • Sterile (aseptic assembly) and certified non-pyrogenic
  • Individual alphanumeric codes for well identification
  • Bulk packed 20 per bag
These 384 well plates are compatible with leading readers and integrate into high-throughput applications.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)