Key Documents
CLS3843
Corning® 96 Well Poly-D-Lysine Treated Microplate
flat bottom clear, white polystyrene, bag of 20, sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear wells
flat bottom clear
round wells
white polystyrene
size:
96 wells
material
clear wells
flat bottom clear
round wells
white polystyrene
sterility
sterile
feature
lid
skirt
packaging
bag of 20
manufacturer/tradename
Corning 3843
size
96 wells
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
poly-D-lysine coated surface
General description
  • Corning® 96 Well Poly-D-Lysine Treated Microplate White walled plates fat bottoms has opaque walls to prevent well-to-well crosstalk
  • 360 μL total volume, Recommended working volumes of 75 to 200 μL.
  • Enhances luminescent signals and has low background luminescence and fluorescence.
  • Optically clear flat well bottom permits direct microscopic viewing.
  • Applicable for both top and bottom reading instruments.
  • Enhanced cell attachment.
  • Nonreversible lids with condensation ring to reduce contamination.
  • Sterile and nonpyrogenic.
  • Individual alphanumeric codes for better identification.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of