Key Documents
CLS3828
Corning® 384 well microplate
Multicoated (Collagen I, Fibronectin, Gelatin, Laminin, Poly-D-Lysine, TC-Treated) surface treatment, black polystyrene, flat bottom, non-sterile, lid, pack of 10
Synonym(s):
3828, multicoated
About This Item
UNSPSC Code:
41121800
material:
black polystyrene
flat bottom
square wells
size:
384 wells
material
black polystyrene
flat bottom
square wells
sterility
non-sterile
feature
lid
Multicoated (Collagen I, Fibronectin, Gelatin, Laminin, Poly-D-Lysine, TC-Treated) surface treatment
packaging
pack of 10
manufacturer/tradename
Corning 3828
size
384 wells
well volume
112 μL
well working volume
20-80 μL
suitability
suitable for tissue
General description
  • Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Individual alphanumeric codes for well identification
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Bui Thi Buu Hue et al.
ChemMedChem, 15(15), 1453-1463 (2020-04-14)
We have synthesized 50 benzimidazole (BMZ) derivatives with 1,2-phenylenediamines and aromatic aldehydes under mild oxidation conditions by using inexpensive, nontoxic inorganic salt sodium metabisulfite in a one-pot condensation reaction and screened their ability to interfere with Zika virus (ZIKV) infection