Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3825BC

Corning® 1536 well plate, with generic barcode

size 1536 wells, flat bottom clear polystyrene wells, non-treated (medium bind), non-sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
material:
flat bottom clear polystyrene wells
polystyrene plate
size:
1536 wells
Pricing and availability is not currently available.

material

flat bottom clear polystyrene wells
polystyrene plate

sterility

non-sterile

feature

non-treated (medium bind)

packaging

pack of 10
case of 50 ea
case of 50

manufacturer/tradename

Corning 3825

maximum volume

12.5 μL

size

1536 wells

working volume

8 μL

suitability

suitable for tissue

Looking for similar products? Visit Product Comparison Guide

General description

Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")
Generic barcoded Corning 1536 well microplates are now available. Code 128 bar code format ensures compatibility with most bar code scanning and software systems. To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code.

  • Resistant to temperature ranges of -80ºC to +52ºC
  • 2.8 mm distance between well bottoms to plate bottom
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface

Microplate Features
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness -minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
  • Working volume of up to 10 μL


Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service