Key Documents
CLS3787
Corning® microcarriers
positive charge, weight 10 g
About This Item
UNSPSC Code:
41122100
feature
positive charge
packaging
pack of 1 ea
manufacturer/tradename
Corning 3787
weight
10 g
General description
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
  • USP Class VI polystyrene material provides a consistent platform
  • Level (SAL) of 10-6
  • Certified nonpyrogenic
These microcarriers are surface treated with a positive charge to enhance cell attachment and expansion.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)