Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3720

Corning® 96 Well Special Optics Microplate

flat bottom clear, black polystyrene, bag of 5, sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, optically clear flat bottom plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

black polystyrene
black/clear bottom polystyrene plate
flat bottom clear

sterility

sterile

feature

lid: no
lid: no
plate format: 96 well special optics

packaging

case of 25
bag of 5

manufacturer/tradename

Corning 3720

maximum volume

360 μL

size

96 wells

working volume

200 μL

color

black

suitability

suitable for (optically clear flat well bottom permits direct microscopic viewing)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 96 Well Special Optics Plates

The 96 well special optics plates have black, opaque walls, and ultra thin flat clear bottoms that permit direct microscopic viewing and improved optical performance in fluorescent-based assays.
  • Ultra thin, optically clear flat well bottom permits direct microscopic viewing, resulting in lower background fluorescence and enabling readings down to 340nm
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 360μL total volume
  • Recommended working volumes of 75 to 200μL
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Without lids

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service