Key Documents
CLS3712
Corning® 384 well plates
black, lid, sterile; γ-irradiated
About This Item
UNSPSC Code:
41100000
material
clear wells
polystyrene
square wells
description
112 μl. black
sterility
sterile; γ-irradiated
feature
lid
packaging
case of 100
manufacturer/tradename
Corning 3712
size
384 wells
well volume
112 μL
well working volume
20-80 μL
color
black
General description
For cell-based fluorescence assays
These 384 well plates are compatible with leading readers and integrate into high-throughput applications.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)