Key Documents
CLS3711
Corning® 384 well microplate
black polystyrene, square, non-sterile, lid: no, case of 100
About This Item
UNSPSC Code:
41100000
Product CLS3711 is not currently sold in your country. Contact Technical Service
material
black polystyrene
square
sterility
non-sterile
feature
lid: no
packaging
case of 100
manufacturer/tradename
Corning 3711
well volume
112 μL
well working volume
20-80 μL
General description
For cell-based fluorescence assays
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1