Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3419

Corning® Transwell® polycarbonate membrane cell culture inserts

75 mm Transwell with 0.4 μm pore polycarbonate membrane insert, TC-treated, w/ lid, sterile, 12/cs

Synonym(s):

cell culture inserts, corning plates, tissue culture plate, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13
Product CLS3419 is not currently sold in your country. Contact Technical Service

material

clear polystyrene (plate or dish)
polycarbonate membrane

sterility

sterile

feature

lid

packaging

pack of 1
case of 12

manufacturer/tradename

Corning 3419

technique(s)

cell culture | mammalian: suitable

insert working volume

9 mL

membrane diam.

75 mm

well working volume

13.0 mL

wells

100 mm , dish

color

translucent

pore size

0.4 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Each insert is packaged in a 100mm dish, 12 inserts per case
  • Membrane must be stained for cell visibility
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines. They feature a 10 μm thin transparent polycarbonate membrane available in a variety of pore sizes. Transwell are treated for optimal cell attachment. Cells must be stained for cell visibility. The PC membrane is compatible with most organic fixatives and stains.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daria Shishkova et al.
International journal of molecular sciences, 21(21) (2020-11-01)
Although saphenous veins (SVs) are commonly used as conduits for coronary artery bypass grafting (CABG), internal thoracic artery (ITA) grafts have significantly higher long-term patency. As SVs and ITA endothelial cells (ECs) have a considerable level of heterogeneity, we suggested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service