Key Documents
CLS3375
Corning® Costar® cell culture flasks
surface area 75 cm2, phenolic-style cap, canted neck
About This Item
UNSPSC Code:
41121804
material
canted neck
polystyrene
sterility
sterile; γ-irradiated
packaging
case of 20 sleeves
sleeve of 5
manufacturer/tradename
Corning
surface area
75 cm2
suitability
suitable for (cell culture applications; optimized for adherent cells)
General description
Corning® Costar® Cell Culture Flasks
  • Disposable
  • Manufactured from optically clear virgin polystyrene with frosted labeling surfaces
  • Volume graduations and alphanumeric coordinates for cell counting
  • Treated for optimal cell attachment
  • Sterilized by gamma irradiation and certified nonpyrogenic
  • 100% integrity tested
Legal Information
Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)