Key Documents
CLS3287
Corning® E-Cube fittings kit
complete fittings kit, 1/cs
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
material
plastic
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3287
General description
E-Cube Fittings Kit contains:
  • 3/16 inch Norprene® pump tubing (18 inches)
  • Tubing ratchet clamps (x6)
  • Barbed T-fittings (x2)
  • Barbed reducing connector (x1)
  • 4 inch wire ties (x20)
  • Barbed Y-connector (x1)
  • Barbed 50 mm PTFE syringe filters (x2)
  • Male barbed luer fittings (x5)
  • Female barbed luer fittings (x5)
  • Male luer plugs (x5)
  • Interlink injection sites (x2)
  • Blunt plastic cannulas (x1)
  • 1.0 inch I.D. × 1.25 inch O.D., 1/8 inch thickness O-rings (x3)
Legal Information
Corning is a registered trademark of Corning, Inc.
E-Cube is a trademark of Corning, Inc.
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)