CLS3150
Corning® Costar® cell culture flasks
162 cm2 traditional straight neck cell culture flask w/ phenolic-style cap, PS, TC-treated, sterile, 25/cs
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
Recommended Products
material
cap (phenolic-style)
phenolic cap
polystyrene
rectangular bottom flask
rectangular flask
straight neck flask
sterility
sterile; γ-irradiated
feature
cap
graduations
packaging
case of 25
pack of 5
manufacturer/tradename
Corning 3150
maximum volume
175 mL
surface area
162 cm2
working volume
32-48 mL
suitability
suitable for (cell culture applications; optimized for adherent cells)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Disposable
- Manufactured from optically clear virgin polystyrene with frosted labeling surfaces
- Volume graduations and alphanumeric coordinates for cell counting
- Treated for optimal cell attachment
- Sterilized by gamma irradiation and certified nonpyrogenic
- 100% integrity tested
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service