CLS3001
Corning® Costar® cell culture flasks
225 cm2 rectangular canted neck cell culture flask w/ vent cap, PS, TC-treated, sterile, 24/cs
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
Recommended Products
material
canted neck flask
cap (vent)
polystyrene
rectangular bottom flask
rectangular flask
vent cap
sterility
sterile; γ-irradiated
feature
cap
graduations
packaging
case of 24
pack of 4
manufacturer/tradename
Corning 3001
maximum volume
370 mL
surface area
225 cm2
working volume
45-67.5 mL
suitability
suitable for (cell culture applications; optimized for adherent cells)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Disposable
- Manufactured from optically clear virgin polystyrene with frosted labeling surfaces
- Volume graduations and alphanumeric coordinates for cell counting
- Treated for optimal cell attachment
- Sterilized by gamma irradiation and certified nonpyrogenic
- 100% integrity tested
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service