Key Documents
CLS2679318
Corning® polypropylene closure
fits neck O.D. 18mm
, clear
About This Item
material
clear
polypropylene
packaging
case of 100
General description
For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)