CLS2509
Corning® 96 Well Sulfhydryl-BIND™ 96 Microplate
flat bottom clear, polystyrene plate, individually wrapped, non-sterile, lid: no
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
About This Item
polystyrene
polystyrene plate
round wells
Recommended Products
material
flat bottom clear
polystyrene
polystyrene plate
round wells
sterility
non-sterile
non-sterile
feature
individually wrapped
lid: no
lid: no
skirt
plate format: 96 well standard
packaging
pack of 1
case of 50
manufacturer/tradename
Corning 2509
size
96 wells
well volume
300 μL
well working volume
75-200 μL
color
clear
suitability
suitable for ELISA applications
binding type
Sulfhydryl-BIND surface
Looking for similar products? Visit Product Comparison Guide
General description
- Sulfhydryl (Sulfhydril-BIND™) binding surface has covalently-linked maleimide groups that covalently couple to sulfhydryl groups via SH moieties
- Suitable for assays requiring site-directed orientation of a biomolecule, especially antibodies
- Without lids, lids supplied separately
- Recommended working volume of 75 to 200 μL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service