CLS2507
Corning® Carbo-BIND™ 96 well plates
96 well plate, Carbo-BIND, polystyrene, flat bottom, clear, non-sterile, 50/cs
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
flat bottom clear wells
polystyrene
round wells
Recommended Products
material
clear polystyrene plate
flat bottom clear wells
polystyrene
round wells
sterility
non-sterile
feature
lid: no
skirt
plate format: 96 well standard
packaging
pack of 1
case of 50
manufacturer/tradename
Corning 2507
size
96 wells
well volume
360 μL
well working volume
75-200 μL
color
clear
suitability
suitable for (assays requiring site-directed orientation of a particular biomolecule)
binding type
Carbo-BIND surface
Looking for similar products? Visit Product Comparison Guide
General description
- Carbohydrate (Carbo-BIND™) binding surface has hydrazide groups covalently coupled to carbohydrate groups
- Ideal for assays requiring site-directed orientation of a biomolecule (oxidized antibodies, carbohydrates, and glycosylated proteins) while maintaining enzymatic or immunological activity
- Without lids, lids supplied separately
- Recommended working volume of 75 to 200 μL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service