Key Documents
CLS2505
Corning® DNA-BIND® 96 well plates
96 well plate, DNA-BIND, polystyrene, flat bottom, nonsterile, clear, 50/cs, non-sterile
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, DNA binding plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear bottom
polystyrene
size:
96 wells
material
clear bottom
polystyrene
sterility
non-sterile
non-sterile
feature
lid: no
skirt
individually wrapped without lid
plate format: standard
well: flat bottom, clear
packaging
pack of 1 (individually wrapped)
case of 50
manufacturer/tradename
Corning 2505
size
96 wells
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (hybridization assays, amplification and other DNA-based assays)
binding type
DNA-BIND® surface
General description
The Corning DNA-BIND surface covalently couples to aminated ssDNA by either the 5′ or 3′ end, making these 96 well plates suitable for hybridization assays, amplification and other DNA-based assays. These polystyrene microplates have a total well volume of 360 μL and a recommended working volume of 75 to 200 μL. Lids are not included.
  • 96 well polystyrene microplates
  • DNA-BIND surface covalently binds to amine groups,providing a convenient method to covalently immobilize aminated ssDNA by either the 5′ or 3′ end.
  • Suitable for hybridization, amplification and for other DNA-based assays
  • Black plates have low background fluorescence and minimize light scattering
  • White plates enhance luminescent signals and have low background luminescence and fluorescence
  • Total well volume: 360 μL; recommended working volumes of 75 to 200 μL
  • Lids are not available
  • Individually wrapped plates
Legal Information
Corning is a registered trademark of Corning, Inc.
DNA-BIND is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)