Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS2498

Corning® DNA-BIND® 96 well plates

96 well plate, DNA-BIND, polystyrene, flat bottom, sterile, black, 50/cs

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, DNA binding plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
material:
black wells
polystyrene
round wells
size:
96 wells

material

black wells
polystyrene
round wells

sterility

non-sterile

feature

lid: no
individually wrapped without lid
plate format: standard
well: flat bottom, black

packaging

pack of 1 (individually wrapped)
case of 50

manufacturer/tradename

Corning 2498

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

black

suitability

suitable for (hybridization assays, amplification and other DNA-based assays)

Looking for similar products? Visit Product Comparison Guide

General description

The Corning DNA-BIND surface covalently couples to aminated ssDNA by either the 5′ or 3′ end, making these 96 well plates suitable for hybridization assays, amplification and other DNA-based assays. These polystyrene microplates have a total well volume of 360 μL and a recommended working volume of 75 to 200 μL. Lids are not included.

  • 96 well polystyrene microplates
  • DNA-BIND surface covalently binds to amine groups,providing a convenient method to covalently immobilize aminated ssDNA by either the 5′ or 3′ end.
  • Suitable for hybridization, amplification and for other DNA-based assays
  • Black plates have low background fluorescence and minimize light scattering
  • White plates enhance luminescent signals and have low background luminescence and fluorescence
  • Total well volume: 360 μL; recommended working volumes of 75 to 200 μL
  • Lids are not available
  • Individually wrapped plates

Legal Information

Corning is a registered trademark of Corning, Inc.
DNA-BIND is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service