CLLS1133
DLD1 CELLS AKT2 -/-
human male colon (Source Disease: Colorectal adenocarcinoma)
Select a Size
$4,530.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$4,530.00
Notify Me
Get notified when this item is ready to ship via email.
General description
This product corresponds to ATCC Cat. No. CCL-221.
Application
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture ratio: approx. 1:3 to 1:10.
The base medium for this cell line is RPMI, Cat. No. R5886. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, L-glutamine, Cat. No. G7513, to a final concentration of 2 mM and sodium pyruvate, Cat. No. S8636, to a final concentration of 1 mM.
Cell freezing medium-DMSO 1X, Cat. No. C6164.
Features and Benefits
Zinc finger nuclease (ZFN) knock out on chromosome 19q13.1-q13.2
Quality
Disclaimer
Kit Components Only
- DLD1 CELLS AKT2 -/-
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service