Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Hepa-T1

99072813, tilapia liver, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

Product Name

Hepa-T1, 99072813

biological source

tilapia liver

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Eel liver

Cell Line Description

Hepa-T1 was derived from normal liver of the fish Tilapia (Oreocheomis niloticus). The cells were described to undergo apoptosis on removal of FBS

Culture Medium

Xcell300 + 2mM Glutamine + 5% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 using 0.25% trypsin or trypsin/EDTA; 26-28°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service