Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Lu6

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 99043004-DNA-5UG
pkg of vial of cells 99043004-1VL

ImmunoDon

(C57BL/6 x BALB/c)F1

Myeloma

P3 x 63 Ag8.653

Reactivity

Anti-HLA DRII

technique(s)

cell culture | mammalian: suitable

isotype

IgG3

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

Lu6 was established from a (C57BL/6 X BALB/c) F1 mouse immunised with a i.p. injection of the lymphoblastoid cell line Lutz Cor and B-pertussis adjuvant. The B cell line was expressing HLA A3, 2; B18, 5; C5; DR3,II. Mice spleen was removed 3 days after immunisation and fused with the myeloma P3x63Ag8.653. Lu6 monoclonal antibodies were shown to react with HLA DRII. Lu6 is also known as LC4A6. It has been described in the International Histocompatibility Workshop II (IIW0912) and 12 (12W539). The cells have been eradicated from mycoplasma using Cibrobay before being deposited at ECACC.

Immunogen

B cell line Lutz Cor HLA A3, 2; B18,5; C5; DR3,II

Application

Tumour studies

Culture Medium

RPMI 1640 + 2 mM glutamine + 1mM sodium pyruvate + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service