Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16-23

Reactivity: Anti-HLA-DR3 and DR52a, 99043001

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

P3x63Ag8.653

Reactivity

Anti-HLA-DR3 and DR52a

technique(s)

cell culture | mammalian: suitable

isotype

IgG3

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The cytotoxic monoclonal antibody 16.23 was obtained from a BALB/c mouse immunised with a single i.p. injection of the human melanoma cell line Mel Juso. Mouse spleen cells were removed 3 days after immunisation and fused with the myeloma P3x63Ag8.653. The Juso B cell line was typed as HLA A1,2; B7,8; CW- ; DR2,3. The monoclonal antibody 16.23 detects the conformation-dependant DR3 epitope but was also reported to react with some HLA-Dw/DRw6 individuals. Together with antibodies S2 (Sigma Catalogue number. 99043002) and S5 (Sigma Catalogue number. 99061827) a useful tool for the biochemical dissection of DRw6 related epitopes has been establised. It has been analysed in the 9th International Histocompatibility Workshop (9W929). The hybridoma line has been eradicated from mycoplasma using Cibrobay before depositing at ECACC.

Immunogen

Human melanoma cell line Mel Juso

Application

Immunohistochemistry, Cytotoxicity, Immunoprecipitation

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM NaP + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service