Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC IAH-CC171

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 99011340-DNA-5UG
pkg of vial of cells 99011340-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS-1

Reactivity

Anti-CD45, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The hybridoma line IAH-CC171 was produced from mice injected with the murine-bovine heterohybrid cell line HBL6 known to express bovine L-CA. Molecules of 180, 205 and 220 kDa are recognised by this antibody. Immunohistology studies showed no reactivity with non-lymphoid cells.

Immunogen

CCI positive heterohybrid cells

Application

Immunohistology, Immunoprecipation, Flow cytometry

Culture Medium

RPMI 1640 + 2 mM glutamine + HT supplement + 10% FBS

Subculture Routine

Maintain cultures between 3-9x 100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service