Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC IAH-CC98

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 99011337-DNA-5UG
pkg of vial of cells 99011337-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS-1

Reactivity

Anti-WC6, bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The monoclonal antibody IAH-CC 98 is part of a cluster known as WC6 that immunoprecipitated a molecule of 210 kDa. This molecule is present on all CD2+ cells and B cells, but not expressed by WC1+cells. Cells have been grown on mouse macrophage feeder cells using hypoxanthine and thymidine at concentrations appropriate for hybridomas. It is recommended to continue this until sufficient cells have been frozen.

Immunogen

Bovine thymocytes

Application

Immunohistology, Flow cytometry, Immunoprecipitation

Culture Medium

RPMI 1640 + 2mM glutamine + HT supplement + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service