LB316
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human blood
packaging
tube of 5 μg 98062333-DNA-5UG
pkg of vial of cells 98062333-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human lymphoblastoid, Xeroderma pigmentosum, EBV-transformed
Cell Line Description
Established by EBV transformation of mononuclear cells from the peripheral blood of a 1 year old male. The patient was diagnosed with Xeroderma pigmentosum, complementation group D, with some features of Cockayne syndrome. Fibroblasts derived from the same individual are designated XP8BR.
Application
Toxicity studies, hprt mutation analysis, DNA repair deficiencies
Culture Medium
RPMI 1640 + 4mM Glutamine + 200 μg/ml Sodium Pyruvate (NaPy) + 15% Heat Inactivated (HI-FBS).
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells/ml, dilute every 1-2 days to 3 x 100,000 cells/ml; 5% CO2; 37°C. Cells form floating aggregates.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service