Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LB750

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human blood

packaging

tube of 5 μg 98062323-DNA-5UG
pkg of vial of cells 98062323-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphoblastoid, trichothiodystrophy, EBV-transformed

Cell Line Description

Established by EBV transformation of mononuclear cells from the peripheral blood from a 10 year old female patient with trichothiodystrophy.

Application

DNA repair deficiency studies, toxicity assays

Culture Medium

RPMI 1640 + 4mM Glutamine + 200 μg/ml Sodium Pyruvate (NaPy) + 15% Heat Inactivated (HI-FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml, dilute every 1-2 days to 3 x 100,000 cells/ml; 5% CO2, 37°C. Cells form floating aggregates.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service