Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55-6

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 97040308-DNA-5UG
pkg of vial of cells 97040308-1VL

ImmunoDon

Mouse BALB/c spleen

Myeloma

Sp2/0-Ag14

Reactivity

Anti-HIV-1 gp120

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma

Cell Line Description

The hybridoma cell line 55-6 produces antibodies reacting with the Human Immunodeficiency Virus I (HIV-1) envelope protein, gp120. Spleen cells from BALB/c mice that had been sequentially immunised with recombinant gp120(rgp120) from three different nonglycosylated isolates of HIV-1 (IIIB,SF2 and Z6) have been fused with Sp2/0-Ag14 myeloma cells. The antibody binds rgp120 from the III B isolate and reacts weakly with gp120 from the MN strain, both shown by ELISA. No cross blocking of any other antibodies in the developed panel was detected, thus indicating that 55-6 binds to a unique site on gp120IIIB. In peptide recognition assays it was shown to bind a synthetic peptide 105-117 (in the CI region) of gp120.

Immunogen

rgp 120

Application

gp120 assays

Culture Medium

RPMI 1640 + 2mM glutamine + HT + 10% FBS

Subculture Routine

Maintain cultures between 1-9 x 100,000 cells/ml; 5% CO2;37ºC

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service