Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CAR-2/3F4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 97011602-DNA-5UG
pkg of vial of cells 97011602-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-Thioredoxin, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

BALB/c mouse spleen x Mouse P3X63Ag8.653

Cell Line Description

CAR-2/3F4, also known as 2/3 F4, was formed by fusing P3X63Ag8-653 mouse myeloma cells with spleen cells from a BALB/c mouse immunised six times with 20μg human thioredoxin (TRX) at monthly intervals. Human TRX was cross-linked to Keyhole Limpet Haemocyanin for this purpose. The antibody is directed against monomeric and polymeric forms of human TRX and shows no cross-reactivity to bacterial TRX or human glutaredoxin. CAR-2/3F4 has been applied as capture antibody in a chemiluminescent detection assay for free human TRX. CAR-XP10 (Sigma Catalogue number. 97011601) can be used as second antibody in this assay.

Immunogen

Human Thioredoxin cross linked to Keyhole Limpet Haemocyanin

Application

Immunohistochemistry, Western blots

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 15% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-9 x 100,000 cells / ml; 5% CO2; 37°C; start culture on mouse macrophage feeder layer

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service