Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OX-86 from rat

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Mouse Hybridoma

packaging

tube of 5 μg 96110601-DNA-5UG
pkg of vial of cells 96110601-1VL

ImmunoDon

Rat AO spleen

Myeloma

NS1

Reactivity

Anti-CD134, mouse

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat AO spleen x Mouse NS1

Cell Line Description

OX-86 was raised by immunising AO rats with recombinant mouse OX-40 antigen and subsequent fusion of spleen cells with NS1 mouse myeloma cells. OX-86 mAb can be used to examine the distribution of OX-40,CD134, a member of the tumour necrosis factor receptor/nerve growth factor receptor super family.

Immunogen

Recombinant mouse OX-40 antigen

Application

OX-40 detection assay
The isotype of the monoclonal antibody produced by this cell line is rat IgG1.

Culture Medium

RPMI 1640 + 2mM glutamine + 10-20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 5x10,000 to 5x100,000 cells/ml: 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service