Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J.CaM1.6

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human blood

packaging

tube of 5 μg 96060401-DNA-5UG
pkg of vial of cells 96060401-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T cell, mutant derivative of Jurkat

Cell Line Description

The T cell leukaemia line J.CaM1.6 was derived from the parent cell line Jurkat E.6-1 (Sigma Catalogue number. 88042803). Jurkat cells were mutagenised with ethyl methanesulfonate and subsequently grown in phytohemagglutinin containing media for 2 weeks. J.CaM1.6 express a structurally normal but functionally defective CD3/Ti complex. The cells fail to exhibit inositolphospholipid metabolism or Ca2+ mobilisation in response to anti-CD3 or anti-Ti monoclonal antibodies. They display resistance to anti-CD3 induced apoptosis, constitutively express Fas and are sensitive to anti-Fas induced apoptosis. It was shown that J.CaM1.6 cells are defective in early signalling events that follow TCR stimulation owing to loss of lck tyrosine kinase function due to the lack of exon 7 in the lck transcript.

Application

Study of apoptosis, signal transduction receptor analysis.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11
D13S317: 8,12
D16S539: 10,11
D5S818: 9
D7S820: 8,10
THO1: 6,9.3
TPOX: 8,10
vWA: 18,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

To keep the cells in exponential growth, maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. If starting from a frozen ampoule, add thawed cells to a conical based centrifuge tube e.g. 15ml tube. Slowly add 4 ml of culture medium to the tube. T

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service