Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMT6/CPR

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse unknown/unspecified

packaging

tube of 5 μg 96042318-DNA-5UG
pkg of vial of cells 96042318-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mammary, drug-resistant

Cell Line Description

The cisplatin-resistant variant of the mouse mammary tumour cell line EMT6/P (Sigma Catalogue number. 96042344) was derived by continuous in vitro exposure to increasing concentrations of cisplatin over a period of 6 weeks. EMT6/CPR was shown to express 2.5-fold resistance to cisplatin as compared with the sensitive parent line. Development of drug resistance was accompanied by a 1.7-fold increase in cellular glutathione (GSH). Growth for 4 weeks in the absence of cisplatin was reported to lead to loss of resistance phenotype and elevated GSH levels in the resistant cell line EMT6/CPR.

Application

Drug-resistance studies

Culture Medium

MEM(EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 0.25μg/ml, cisplatin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37ºC. It is recommended to culture the cells without drugs upon resuscitation until the first passage.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service