Hypertriploid. The modal chromosome number was 80, occurring in 42% of cells. Cells with 78 chromosomes also occurred at a high frequency.
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Bladder Transitional Cell Carcinoma
Cell Line Description
Derived from a urinary bladder transitional cell carcinoma from a human male. These cells are tumourigenic in nude mice when 10E7 cells are inoculated subcutaneously. Hypertriploid. The modal chromosome number was 80, occurring in 42% of cells. Cells with 78 chromosomes also occurred at a high frequency. The rate of cells with higher ploidies was 2.5%. There were 30 or more marker chromosomes in each cell. They included der(1)t (1;?) (p32;?), ?t(1p5p), i(3q), t(7q14q), ?t(2p3p) and others. The X and N3 had single copy per cell, and others were generally two or three copies per cell.
Split sub-confluent cultures (70-80 %) 1:4 to 1:10 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 37 C; 5% CO2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.