Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ED7

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 95110624-DNA-5UG
pkg of vial of cells 95110624-1VL

ImmunoDon

Mouse spleen cells

Myeloma

Sp2/0

Reactivity

Anti-CD11b (CR3), rat

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse spleen

Cell Line Description

The ED7 hybridoma secretes antibodies reactive against rat monocytes, macrophages and granulocytes. Two proteins of 160kDa and 95kDa are immunoprecipitated under reducing conditions. The antibody also recognises rat SIRP (personal communication).

Immunogen

Wistar rat spleen

Application

Immunohisto-and cytochemistry, FACS

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service