Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-tyr-1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing

Select a Size

95092903-DNA-5UG
$402.00
1 VIAL
$981.00

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery


Select a Size

Change View
95092903-DNA-5UG
$402.00
1 VIAL
$981.00

About This Item

UNSPSC Code:
41106514

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 95092903-DNA-5UG
pkg of vial of cells 95092903-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-Phosphotyrosine

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8.653 x Mouse BALB/c spleen

Cell Line Description

The hybridoma cell line P-tyr-1 was produced by fusing P3X63Ag8.653 mouse myeloma cells with spleen cells from a female BALB/c mouse immunised with aminobenzyl phosphonate. Aminobenzyl phosphonate is a structural analogue of the side-chain in phosphotyrosine, and was coupled to hemocyanin (antibody P-HC) for immunisation. The secreted IgG1 antibodies react with phosphotyrosine and will detect phosphotyrosine-containing proteins by immunoprecipitation and Western blot analysis. The antibody does not react with phosphoserine or phosphothreonine. P-tyr-1 was tested for Bovine Viral Diarrhoea Virus (BVDV) at ECACC and found to be negative.

Immunogen

Aminobenzyl phosphonate

Application

Detection of phosphotyrosine-containing proteins

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service