Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WR19L

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95050414, mouse lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse lymph

packaging

tube of 5 μg 95050414-DNA-5UG
pkg of vial of cells 95050414-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Moloney murine leukemia virus

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse lymphoma

Cell Line Description

The WR19L cell line was derived from the WR19 tumour induced by Abelson murine leukemia virus but does not produce Abelson virus. Mouse strain: BALB/c. Antigen expression: H-2d; Thy-1.2

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Seed cells at 2 x 105 cells/ml. Maintain cultures between 1 x 105 to 1 x 106 cells/ml; 5% CO2; 37°C. Split ratio: 1:2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service